siRNA Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134300.2 | NP_001127772.1 | 4 | 878 | ||
| GenBank mRNA | AK029844.1 | - | 6 | 1357 | ||
| AK147524.1 | - | 4 | 880 | |||
| BC075648.1 | - | 3 | 289 | 30620653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nol4l| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134300.2 | NP_001127772.1 | 4 | 878 | ||
| GenBank mRNA | AK029844.1 | - | 6 | 1357 | ||
| AK147524.1 | - | 4 | 880 | |||
| BC075648.1 | - | 3 | 289 | 30620653 | ||