siRNA Details
| Gene Symbol: Gm826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033411.3 | NP_001028583.1 | 1, 2 | 387 | ||
| GenBank mRNA | AK142710.1 | - | 1, 2 | 388 | ||
| BC147270.1 | - | 1, 2 | 335 | 9056346 | ||
| BC147271.1 | - | 1, 2 | 335 | 9056326 | ||

Gm826
Genomic Map