siRNA Details
| Gene Symbol: Sertm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177854.4 | NP_808522.1 | 3 | 670 | ||
| GenBank mRNA | AK031306.1 | - | 3 | 671 | ||
| BC058995.1 | - | 2 | 483 | 5708109 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sertm1| Gene Symbol: Sertm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177854.4 | NP_808522.1 | 3 | 670 | ||
| GenBank mRNA | AK031306.1 | - | 3 | 671 | ||
| BC058995.1 | - | 2 | 483 | 5708109 | ||