siRNA Details
| Gene Symbol: Fnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162999.2 | NP_001156471.1 | 9 | 1106 | ||
| GenBank mRNA | AK122512.1 | - | 1 | 183 | ||
| BC116316.1 | - | 2 | 370 | |||
| BC116317.1 | - | 2 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fnip2| Gene Symbol: Fnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162999.2 | NP_001156471.1 | 9 | 1106 | ||
| GenBank mRNA | AK122512.1 | - | 1 | 183 | ||
| BC116316.1 | - | 2 | 370 | |||
| BC116317.1 | - | 2 | 370 | |||