siRNA Details
| Gene Symbol: Fam166b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162381.1 | NP_001155853.1 | 2 | 238 | ||
| NM_177377.5 | NP_796351.2 | 2 | 238 | |||
| GenBank mRNA | AK029435.1 | - | 2 | 239 | ||
| AK053208.1 | - | 2 | 239 | |||
| BC172030.1 | - | 2 | 221 | |||

Fam166b
Genomic Map