siRNA Details
| Gene Symbol: Tmem61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028711.1 | - | 2 | 999 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem61| Gene Symbol: Tmem61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028711.1 | - | 2 | 999 | ||