siRNA Details
| Gene Symbol: Skint2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285963.1 | NP_001272892.1 | 5 | 767 | ||
| NM_001285965.1 | NP_001272894.1 | 5 | 767 | |||
| GenBank mRNA | AK040156.1 | - | 4 | 756 | ||
| EF494892.1 | - | 5 | 767 | |||
| EF494893.1 | - | 4 | 767 | |||
| EU099299.1 | - | 5 | 767 | |||
| EU099300.1 | - | 5 | 767 | |||
| EU650234.1 | - | 4 | 741 | |||

Skint2
Genomic Map