siRNA Details
| Gene Symbol: Cxcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203320.3 | NP_976065.1 | 2 | 222 | ||
| GenBank mRNA | AK144158.1 | - | 2 | 225 | ||
| AK150314.1 | - | 2 | 221 | |||
| AK154334.1 | - | 2 | 221 | |||
| AK155702.1 | - | 313 | ||||
| AY311403.1 | - | 2 | 220 | |||
| BC117014.1 | - | 2 | 177 | 40126333 | ||
| BC117016.1 | - | 2 | 177 | 40126335 | ||

Cxcl3
Genomic Map