siRNA Details
| Gene Symbol: Hfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177873.3 | NP_808541.2 | 35 | 3959 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hfm1| Gene Symbol: Hfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177873.3 | NP_808541.2 | 35 | 3959 | ||