siRNA Details
| Gene Symbol: Hfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252516.1 | NP_001239445.1 | 9 | 1124 | ||
| NM_177873.3 | NP_808541.2 | 9 | 1198 | |||
| GenBank mRNA | AK039266.1 | - | 9 | 1125 | ||
| BC094509.1 | - | 4 | 339 | |||
| BC172050.1 | - | 8 | 1099 | |||
| BC172051.1 | - | 9 | 1160 | |||
| BC172061.1 | - | 9 | 1160 | 9054347 | ||

Hfm1
Genomic Map