siRNA Details
| Gene Symbol: Taok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081308.2 | NP_001074777.1 | 15 | 1912 | ||
| NM_001199685.1 | NP_001186614.1 | 16 | 2020 | |||
| NM_183306.2 | NP_899129.2 | 16 | 1997 | |||
| GenBank mRNA | AK039665.1 | - | 3 | 570 | ||
| AK042181.1 | - | 16 | 1920 | |||
| AK155208.1 | - | 15 | 1908 | |||
| AK188887.1 | - | 1 | 51 | |||
| BC158072.1 | - | 16 | 1962 | 9088167 | ||

Taok3
Genomic Map