siRNA Details
| Gene Symbol: Taok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081308.2 | NP_001074777.1 | 18 | 2595 | ||
| NM_001199685.1 | NP_001186614.1 | 19 | 2703 | |||
| NM_183306.2 | NP_899129.2 | 19 | 2680 | |||
| GenBank mRNA | AK039665.1 | - | 6 | 1253 | ||
| AK217809.1 | - | 2 | 214 | |||
| BC158072.1 | - | 19 | 2645 | 9088167 | ||
| BM225493.2 | - | 3 | 401 | |||

Taok3
Genomic Map