siRNA Details
Gene Symbol: Thsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164805.1 | NP_001158277.1 | 22 | 4363 | ||
GenBank mRNA | AK133324.1 | - | 17 | 2443 | ||
AK173072.1 | - | 22 | 4134 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164805.1 | NP_001158277.1 | 22 | 4363 | ||
GenBank mRNA | AK133324.1 | - | 17 | 2443 | ||
AK173072.1 | - | 22 | 4134 |