siRNA Details
| Gene Symbol: Thsd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164805.1 | NP_001158277.1 | 23 | 4546 | ||
| GenBank mRNA | AK133324.1 | - | 18 | 2626 | ||
| AK173072.1 | - | 23 | 4317 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thsd7a| Gene Symbol: Thsd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164805.1 | NP_001158277.1 | 23 | 4546 | ||
| GenBank mRNA | AK133324.1 | - | 18 | 2626 | ||
| AK173072.1 | - | 23 | 4317 | |||