siRNA Details
| Gene Symbol: Wipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167860.1 | NP_001161332.1 | 4 | 505 | ||
| NM_001167861.1 | NP_001161333.1 | 4 | 505 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wipf3| Gene Symbol: Wipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167860.1 | NP_001161332.1 | 4 | 505 | ||
| NM_001167861.1 | NP_001161333.1 | 4 | 505 | |||