siRNA Details
| Gene Symbol: Far2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178797.3 | NP_848912.1 | 5 | 764 | ||
| GenBank mRNA | AK038584.1 | - | 5 | 717 | ||
| AK043388.1 | - | 5 | 764 | |||
| AK043589.1 | - | 5 | 764 | |||
| AK137298.1 | - | 5 | 764 | |||
| AK161201.1 | - | 5 | 731 | |||
| BC055759.1 | - | 5 | 742 | 6809131 | ||

Far2
Genomic Map