siRNA Details
| Gene Symbol: Ctxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183315.2 | NP_899138.1 | 2 | 255 | ||
| GenBank mRNA | AK154120.1 | - | 2 | 206 | ||
| AK158759.1 | - | 2 | 149 | |||
| BC028881.1 | - | 2 | 191 | 4224851 | ||
| DN176231.1 | - | 2 | 255 | |||

Ctxn1
Genomic Map