siRNA Details
| Gene Symbol: Zfp866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177899.3 | NP_808567.2 | 4 | 1589 | ||
| GenBank mRNA | AK052363.1 | - | 4 | 1568 | ||
| AK137334.1 | - | 4 | 1591 | |||
| AK140357.1 | - | 4 | 1584 | |||
| BC057095.1 | - | 4 | 1563 | 6419770 | ||
| BC117846.1 | - | 4 | 1544 | 40093653 | ||
| BC117847.1 | - | 4 | 1544 | 40093655 | ||
| BC138224.1 | - | 4 | 1540 | 8861243 | ||
| BC138227.1 | - | 4 | 1540 | 8861246 | ||

Zfp866
Genomic Map