siRNA Details
Gene Symbol: Rnf150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177378.4 | NP_796352.2 | 1 | 1038 | ||
GenBank mRNA | AK032722.1 | - | 1 | 1058 | ||
AK041412.1 | - | 1 | 1040 | |||
AK170194.1 | - | 2 | 359 | |||
AK220374.1 | - | 1 | 921 |
