siRNA Details
| Gene Symbol: Rnf150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177378.4 | NP_796352.2 | 1 | 1106 | ||
| GenBank mRNA | AK032722.1 | - | 1 | 1126 | ||
| AK041412.1 | - | 1 | 1108 | |||
| AK170194.1 | - | 2 | 427 | |||
| AK220374.1 | - | 1 | 989 | |||

Rnf150
Genomic Map