siRNA Details
Gene Symbol: Rnf150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177378.4 | NP_796352.2 | 1 | 732 | ||
GenBank mRNA | AK032722.1 | - | 1 | 752 | ||
AK220374.1 | - | 1 | 615 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177378.4 | NP_796352.2 | 1 | 732 | ||
GenBank mRNA | AK032722.1 | - | 1 | 752 | ||
AK220374.1 | - | 1 | 615 |