siRNA Details
Gene Symbol: Dhps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039514.1 | NP_001034603.1 | 8 | 1021 | ||
GenBank mRNA | AK159104.1 | - | 8 | 1022 | ||
BC039963.1 | - | 6 | 807 | 4240646 | ||
BC061234.1 | - | 7 | 860 |
