siRNA Details
| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039514.1 | NP_001034603.1 | 6 | 829 | ||
| GenBank mRNA | AK159104.1 | - | 6 | 830 | ||
| BC039963.1 | - | 4 | 615 | 4240646 | ||
| BC061234.1 | - | 5 | 668 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhps| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039514.1 | NP_001034603.1 | 6 | 829 | ||
| GenBank mRNA | AK159104.1 | - | 6 | 830 | ||
| BC039963.1 | - | 4 | 615 | 4240646 | ||
| BC061234.1 | - | 5 | 668 | |||