siRNA Details
Gene Symbol: Drc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042715.3 | NP_001036180.2 | 16 | 2334 | ||
GenBank mRNA | AY352586.2 | - | 16 | 2331 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Drc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042715.3 | NP_001036180.2 | 16 | 2334 | ||
GenBank mRNA | AY352586.2 | - | 16 | 2331 |