siRNA Details
| Gene Symbol: AI593442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286641.1 | NP_001273570.1 | 2 | 720 | ||
| NM_178906.5 | NP_849237.2 | 2 | 671 | |||
| GenBank mRNA | AK033979.1 | - | 2 | 670 | ||
| BC109328.2 | - | 2 | 540 | 40051267 | ||
| BM933125.1 | - | 1 | 156 | |||

AI593442
Genomic Map