siRNA Details
| Gene Symbol: Slc51b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178933.2 | NP_849264.1 | 2, 3 | 148 | ||
| GenBank mRNA | AK002463.1 | - | 2, 3 | 279 | ||
| AK004188.1 | - | 2, 3 | 331 | |||
| AY279396.1 | - | 2, 3 | 148 | |||
| BC115606.1 | - | 1, 2 | 127 | 40102690 | ||
| BC115607.1 | - | 1, 2 | 127 | 40102691 | ||

Slc51b
Genomic Map