siRNA Details
| Gene Symbol: Gmppb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177910.3 | NP_808578.1 | 2, 3 | 442 | ||
| GenBank mRNA | AK088295.1 | - | 2, 3 | 424 | ||
| AK148125.1 | - | 442 | ||||
| AK158470.1 | - | 2, 3 | 442 | |||
| AK165816.1 | - | 2, 3 | 444 | |||
| BC061207.1 | - | 3, 4 | 316 | 30249330 | ||

Gmppb
Genomic Map