siRNA Details
| Gene Symbol: Gm5124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032684.1 | - | 1 | 133 | ||
| BC137827.1 | - | 1 | 113 | 8860846 | ||
| BM943806.1 | - | 1 | 47 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5124| Gene Symbol: Gm5124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032684.1 | - | 1 | 133 | ||
| BC137827.1 | - | 1 | 113 | 8860846 | ||
| BM943806.1 | - | 1 | 47 | |||