siRNA Details
Gene Symbol: Gm5124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032684.1 | - | 1 | 717 | ||
BC137827.1 | - | 1 | 697 | 8860846 | ||
BM943806.1 | - | 1 | 631 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032684.1 | - | 1 | 717 | ||
BC137827.1 | - | 1 | 697 | 8860846 | ||
BM943806.1 | - | 1 | 631 |