siRNA Details
| Gene Symbol: Zdhhc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007460.1 | NP_001007461.1 | 3 | 938 | ||
| GenBank mRNA | AK137472.1 | - | 2 | 548 | ||
| AK147553.1 | - | 4 | 1299 | |||
| AY668949.1 | - | 3 | 938 | |||
| BC139052.1 | - | 3 | 981 | 8862074 | ||
| BC145525.1 | - | 3 | 972 | 9054088 | ||
| BC145527.1 | - | 3 | 1031 | |||
| BC145684.1 | - | 3 | 981 | 40130987 | ||

Zdhhc23
Genomic Map