siRNA Details
Gene Symbol: Gm853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034872.2 | NP_001030044.1 | 7, 8 | 1115 | ||
GenBank mRNA | AK143920.1 | - | 7, 8 | 1116 | ||
BC147394.1 | - | 6, 7 | 980 | 9056469 | ||
BC147395.1 | - | 6, 7 | 980 | 9056458 | ||
BC172060.1 | - | 6, 7 | 980 | 9054346 |
