siRNA Details
| Gene Symbol: Ksr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114545.2 | NP_001108017.2 | 1, 2 | 1015 | ||
| GenBank mRNA | AK137433.1 | - | 1, 2 | 1017 | ||
| BC150883.1 | - | 1, 2 | 1015 | |||
| BC150890.1 | - | 1, 2 | 1015 | |||
| DQ531035.1 | - | 1, 2 | 188 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006249485.

Ksr2
Genomic Map