siRNA Details
| Gene Symbol: Ksr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114545.2 | NP_001108017.2 | 4 | 1718 | ||
| GenBank mRNA | AK137433.1 | - | 4 | 1720 | ||
| BC068233.1 | - | 2 | 503 | |||
| BC150883.1 | - | 4 | 1718 | |||
| BC150890.1 | - | 4 | 1718 | |||
| DQ531035.1 | - | 4 | 879 | |||

Ksr2
Genomic Map