siRNA Details
| Gene Symbol: Fkbp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045528.1 | NP_001038993.1 | 4, 5 | 453 | ||
| GenBank mRNA | AK049459.1 | - | 4, 5 | 398 | ||
| AK049509.1 | - | 4, 5 | 454 | |||
| AK082565.1 | - | 4, 5 | 402 | |||
| AK085087.1 | - | 4, 5 | 441 | |||
| AK161705.1 | - | 4, 5 | 383 | |||
| BC060651.1 | - | 4, 5 | 367 | |||
| BC137706.1 | - | 4, 5 | 387 | 8860725 | ||

Fkbp15
Genomic Map