siRNA Details
| Gene Symbol: Myo1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177390.3 | NP_796364.2 | 17 | 2413 | ||
| GenBank mRNA | AK049374.1 | - | 17 | 2414 | ||
| AK148112.1 | - | 4 | 532 | |||
| BC034071.1 | - | 3 | 393 | |||
| BC039700.1 | - | 12 | 1545 | |||
| BC150765.1 | - | 17 | 2279 | 9087676 | ||

Myo1d
Genomic Map