siRNA Details
Gene Symbol: Nalcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177393.4 | NP_796367.3 | 15 | 2148 | ||
GenBank mRNA | AK043071.1 | - | 15 | 1846 | ||
AK143219.1 | - | 14 | 1980 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nalcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177393.4 | NP_796367.3 | 15 | 2148 | ||
GenBank mRNA | AK043071.1 | - | 15 | 1846 | ||
AK143219.1 | - | 14 | 1980 |