siRNA Details
| Gene Symbol: Nalcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177393.4 | NP_796367.3 | 3 | 601 | ||
| GenBank mRNA | AK036122.1 | - | 3 | 419 | ||
| AK039034.1 | - | 3 | 419 | |||
| AK040761.1 | - | 3 | 593 | |||
| AK043071.1 | - | 3 | 334 | |||
| AK143219.1 | - | 3 | 601 | |||

Nalcn
Genomic Map