siRNA Details
| Gene Symbol: Dppa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163358.1 | NP_001156830.1 | 6 | 376 | ||
| NM_178247.3 | NP_839978.1 | 7 | 462 | |||
| GenBank mRNA | AF490345.1 | - | 7 | 448 | ||
| AK145642.1 | - | 7 | 463 | |||
| AK166752.1 | - | 7 | 460 | |||
| BC115806.1 | - | 6 | 403 | 40106955 | ||
| BC115901.1 | - | 6 | 400 | 40106956 | ||

Dppa1
Genomic Map