siRNA Details
| Gene Symbol: Pcdha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198117.2 | NP_932785.1 | 1 | 747 | ||
| GenBank mRNA | AY013760.1 | - | 1 | 634 | ||
| BC030897.1 | - | 1 | 747 | |||
| BC141390.1 | - | 1 | 728 | 9056003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha2| Gene Symbol: Pcdha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198117.2 | NP_932785.1 | 1 | 747 | ||
| GenBank mRNA | AY013760.1 | - | 1 | 634 | ||
| BC030897.1 | - | 1 | 747 | |||
| BC141390.1 | - | 1 | 728 | 9056003 | ||