siRNA Details
| Gene Symbol: Clec18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181549.3 | NP_853527.2 | 11 | 1374 | ||
| GenBank mRNA | AK132888.1 | - | 13 | 2242 | ||
| AY223868.1 | - | 1582 | ||||
| AY223869.1 | - | 1553 | ||||
| AY223870.1 | - | 1557 | ||||
| AY223871.1 | - | 1463 | ||||
| AY223872.1 | - | 1528 | ||||
| AY223873.1 | - | 1434 | ||||
| CO044557.1 | - | 1, 2 | 459 | |||

Clec18a
Genomic Map