siRNA Details
Gene Symbol: Gm5136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203660.2 | NP_988991.1 | 1 | 285 | ||
GenBank mRNA | BC120506.1 | - | 1 | 285 | 40129429 | |
BC120508.1 | - | 1 | 285 | 40129431 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203660.2 | NP_988991.1 | 1 | 285 | ||
GenBank mRNA | BC120506.1 | - | 1 | 285 | 40129429 | |
BC120508.1 | - | 1 | 285 | 40129431 |