siRNA Details
| Gene Symbol: Morn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194269.2 | NP_919245.1 | 4 | 403 | ||
| GenBank mRNA | AY367765.1 | - | 4 | 403 | ||
| BC037365.1 | - | 3 | 329 | |||
| BC089486.1 | - | 2 | 178 | 6742814 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morn2| Gene Symbol: Morn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194269.2 | NP_919245.1 | 4 | 403 | ||
| GenBank mRNA | AY367765.1 | - | 4 | 403 | ||
| BC037365.1 | - | 3 | 329 | |||
| BC089486.1 | - | 2 | 178 | 6742814 | ||