siRNA Details
| Gene Symbol: Tlcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027249.3 | NP_081525.1 | 4 | 892 | ||
| GenBank mRNA | BC021945.1 | - | 4 | 863 | 5098319 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlcd2| Gene Symbol: Tlcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027249.3 | NP_081525.1 | 4 | 892 | ||
| GenBank mRNA | BC021945.1 | - | 4 | 863 | 5098319 | |