siRNA Details
| Gene Symbol: Gm884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033434.2 | NP_001028606.2 | 1 | 170 | ||
| GenBank mRNA | AK029868.1 | - | 1 | 170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm884| Gene Symbol: Gm884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033434.2 | NP_001028606.2 | 1 | 170 | ||
| GenBank mRNA | AK029868.1 | - | 1 | 170 | ||