siRNA Details
Gene Symbol: Atxn7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028139.5 | NP_082415.2 | 1 | 216 | ||
GenBank mRNA | AK013145.1 | - | 1 | 123 | ||
BB843098.1 | - | 1 | 139 | |||
BC017546.1 | - | 1 | 86 | |||
BC100598.1 | - | 1 | 97 | |||
BY019350.1 | - | 1 | 209 | |||
BY203850.1 | - | 1 | 88 |
