siRNA Details
| Gene Symbol: Ccdc177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008423.2 | NP_001008423.2 | 2 | 1017 | ||
| GenBank mRNA | AK147473.1 | - | 844 | |||
| BC067044.1 | - | 2 | 989 | 30531780 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK141165.1 | - | 422 | Chr. - - [-] on Build | ||

Ccdc177
Genomic Map