siRNA Details
| Gene Symbol: Zfp729b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163246.2 | NP_001156718.1 | 4 | 3810 | ||
| GenBank mRNA | AK164508.1 | - | 1 | 693 | ||
| AK170565.1 | - | 3 | 2098 | |||
| BC047145.1 | - | 4 | 3766 | 6467898 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp729b| Gene Symbol: Zfp729b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163246.2 | NP_001156718.1 | 4 | 3810 | ||
| GenBank mRNA | AK164508.1 | - | 1 | 693 | ||
| AK170565.1 | - | 3 | 2098 | |||
| BC047145.1 | - | 4 | 3766 | 6467898 | ||