siRNA Details
| Gene Symbol: Gm5142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004158.2 | NP_001004158.1 | 1 | 224 | ||
| GenBank mRNA | AK076969.1 | - | 1 | 225 | ||
| BC099536.1 | - | 1 | 196 | 30918677 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5142| Gene Symbol: Gm5142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004158.2 | NP_001004158.1 | 1 | 224 | ||
| GenBank mRNA | AK076969.1 | - | 1 | 225 | ||
| BC099536.1 | - | 1 | 196 | 30918677 | ||