siRNA Details
| Gene Symbol: Zfp395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199029.2 | NP_950194.2 | 9 | 1467 | ||
| GenBank mRNA | AK155538.1 | - | 9 | 1468 | ||
| CF737135.1 | - | 4 | 561 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp395| Gene Symbol: Zfp395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199029.2 | NP_950194.2 | 9 | 1467 | ||
| GenBank mRNA | AK155538.1 | - | 9 | 1468 | ||
| CF737135.1 | - | 4 | 561 | |||