siRNA Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033439.3 | NP_001028611.2 | 16, 17 | 1900 | ||
| GenBank mRNA | AK129264.1 | - | 16, 17 | 1889 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrch1| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033439.3 | NP_001028611.2 | 16, 17 | 1900 | ||
| GenBank mRNA | AK129264.1 | - | 16, 17 | 1889 | ||